[1,2,4]Triazolo[4,3-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
37921-1G
37921-25G
37921-5G
76889-196EA
62.6
USD
76889-196
76889-198
76889-200
[1,2,4]Triazolo[4,3-a]pyridine ≥97%
[1,2,4]Triazolo[4,3-a]pyridine
Formula:
C₆H₅N₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD09994416 CAS Number: 274-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...