2,3-Difluoro-5-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37735-1G 37735-500MG 37735-5G
76889-150EA 587.74 USD
76889-150 76889-152 76889-154
2,3-Difluoro-5-methoxy-4-methylbenzaldehyde ≥97%
2,3-Difluoro-5-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986838
CAS Number: 1706430-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR