(E)-3-(4-chloro-3,5-dimethylphenyl)acrylic acid ≥95%

Supplier: AOBChem USA

Warning

37627-1G 37627-25G 37627-500MG 37627-5G
76889-108EA 260.99 USD
76889-108 76889-110 76889-112 76889-114
(E)-3-(4-chloro-3,5-dimethylphenyl)acrylic acid ≥95%
(E)-3-(4-chloro-3,5-dimethylphenyl)acrylic acid
Formula: C₁₁H₁₁ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632246
CAS Number: 2387419-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR