(4-Fluoro-2,5-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

37516-1G 37516-25G 37516-5G 37516-500MG
76889-050EA 91.24 USD
76889-050 76889-052 76889-054 77650-456
(4-Fluoro-2,5-dimethylphenyl)boronic acid ≥97%
(4-Fluoro-2,5-dimethylphenyl)boronic acid
Formula: C₈H₁₀BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD10699621
CAS Number: 1072946-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR