2-(Difluoromethoxy)naphthalene ≥97%

Supplier: AOBChem USA

Warning

37494-1G 37494-25G 37494-5G 37494-500MG
76889-036EA 98.67 USD
76889-036 76889-038 76889-040 77650-450
2-(Difluoromethoxy)naphthalene ≥97%
2-(Difluoromethoxy)naphthalene
Formula: C₁₁H₈F₂O
MW: 194.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05856249
CAS Number: 712-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR