2,3-Difluoro-1-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37468-1G 37468-25G 37468-500MG 37468-5G
76889-008EA 379.81 USD
76889-008 76889-010 76889-012 76889-014
2,3-Difluoro-1-iodo-4-methoxybenzene ≥97%
2,3-Difluoro-1-iodo-4-methoxybenzene
Formula: C₇H₅F₂IO
MW: 270.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913391
CAS Number: 156499-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR