1,5-Dibromo-3-fluoro-2-isopropoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

37336-1G 37336-25G 37336-5G
76888-984EA 62.6 USD
76888-984 76888-986 76888-988
1,5-Dibromo-3-fluoro-2-isopropoxybenzene ≥97% research grade
1,5-Dibromo-3-fluoro-2-isopropoxybenzene
Formula: C₉H₉Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD32632200
CAS Number: 2404733-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR