(3-Bromo-2-Methoxyphenyl)(Methyl)Sulfane 95%

Supplier: AOBChem USA

Warning

37314-1G 37314-25G 37314-5G 37314-500MG
76888-972EA 115.64 USD
76888-972 76888-974 76888-976 77650-446
(3-Bromo-2-Methoxyphenyl)(Methyl)Sulfane 95%
(3-Bromo-2-Methoxyphenyl)(Methyl)Sulfane
Formula: C₈H₉BrOS
MW: 233.125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22488894
CAS Number: 1824368-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR