1-(3-Chlorophenyl)-1H-pyrrole ≥97%
Supplier: AOBChem USA
|
Warning
|
37274-1G
37274-25G
37274-5G
76888-950EA
62.6
USD
76888-950
76888-952
76888-954
1-(3-Chlorophenyl)-1H-pyrrole ≥97%
1-(3-Chlorophenyl)-1H-pyrrole
Precaución: for Research Use Only
Formula:
C₁₀H₈ClN MW: 177.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02665247 CAS Number: 41910-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...