1-(4-Methoxyphenyl)-1H-pyrrole ≥97%

Supplier: AOBChem USA

Warning

37268-1G 37268-25G 37268-5G
76888-940EA 70.02 USD
76888-940 76888-942 76888-944
1-(4-Methoxyphenyl)-1H-pyrrole ≥97%
1-(4-Methoxyphenyl)-1H-pyrrole

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD00128350
CAS Number: 5145-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR