(4-Bromo-3-Chloro-2-Methoxyphenyl)(Methyl)Sulfane ≥97% research grade

Supplier: AOBChem USA

Warning

37248-1G 37248-25G 37248-500MG 37248-5G
76888-924EA 379.81 USD
76888-924 76888-926 76888-928 76888-930
(4-Bromo-3-Chloro-2-Methoxyphenyl)(Methyl)Sulfane ≥97% research grade
(4-Bromo-3-Chloro-2-Methoxyphenyl)(Methyl)Sulfane
Formula: C₈H₈BrClOS
Storage Temperature: Refrigerator
MDL Number: MFCD32707017
CAS Number: 2413441-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR