5-Bromo-2,3-difluorocinnamic acid 95%

Supplier: AOBChem USA

Warning

Synonyms: (E)-3-(5-bromo-2,3-difluorophenyl)acrylic acid

37243-1G 37243-500MG 37243-5G 37243-25G
76888-912EA 528.33 USD
76888-912 76888-914 76888-916 77655-466
5-Bromo-2,3-difluorocinnamic acid 95%
5-Bromo-2,3-difluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₅BrF₂O₂
MW: 263.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632174
CAS Number: 2387460-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR