1-(2-Bromophenyl)pyrrole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Bromophenyl)-1H-pyrrole

37075-1G 37075-25G 37075-5G
76888-856EA 66.84 USD
76888-856 76888-858 76888-860
1-(2-Bromophenyl)pyrrole ≥97%
1-(2-Bromophenyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.0800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02665226
CAS Number: 69907-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR