(2-Chloro-5-Methoxyphenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

37035-25G 37035-5G
76888-830EA 453.01 USD
76888-830 76888-832
(2-Chloro-5-Methoxyphenyl)(Methyl)Sulfane ≥97%
(2-Chloro-5-Methoxyphenyl)(Methyl)Sulfane
Formula: C₈H₉ClOS
MW: 188.674 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489297
CAS Number: 473255-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR