1-(Methoxymethyl)-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

36994-1G 36994-25G 36994-5G
76888-804EA 62.6 USD
76888-804 76888-806 76888-808
1-(Methoxymethyl)-3,5-bis(trifluoromethyl)benzene ≥95%
1-(Methoxymethyl)-3,5-bis(trifluoromethyl)benzene
Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632125
CAS Number: 887577-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR