2-Bromo-5-fluoro-p-xylene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-fluoro-2,5-dimethylbenzene

36990-25G 36990-5G
76888-792EA 516.66 USD
76888-792 76888-794
2-Bromo-5-fluoro-p-xylene ≥97% research grade
2-Bromo-5-fluoro-p-xylene
Formula: C₈H₈BrF
MW: 203.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878421
CAS Number: 51760-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR