2,3-Difluoro-4,5-dimethoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

36932-1G 36932-500MG 36932-5G
76888-752EA 658.82 USD
76888-752 76888-754 76888-756
2,3-Difluoro-4,5-dimethoxybenzoic acid ≥97% research grade
2,3-Difluoro-4,5-dimethoxybenzoic acid
Formula: C₉H₈F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD32632114
CAS Number: 2379322-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR