1-(4-Chloro-3-methoxyphenyl)ethanone ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
36929-1G
36929-25G
36929-500MG
36929-5G
76888-738EA
292.81
USD
76888-738
76888-740
76888-742
76888-744
1-(4-Chloro-3-methoxyphenyl)ethanone ≥97% research grade
1-(4-Chloro-3-methoxyphenyl)ethanone
Formula:
C₉H₉ClO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11553430 CAS Number: 73898-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...