1,2-Dichloro-4-fluoro-3-iodobenzene 97%

Supplier: AOBChem USA

Warning

36919-1G 36919-25G 36919-5G 36919-500MG
76888-724EA 101.85 USD
76888-724 76888-726 76888-728 77646-430
1,2-Dichloro-4-fluoro-3-iodobenzene 97%
1,2-Dichloro-4-fluoro-3-iodobenzene
Formula: C₆H₂Cl₂FI
MW: 290.889 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28786112
CAS Number: 1804886-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR