4-Chloro-2-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Chloro-2-methoxyphenyl)methanol

36897-25G 36897-5G
76888-714EA 230.22 USD
76888-714 76888-716
4-Chloro-2-methoxybenzyl alcohol ≥97%
4-Chloro-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD02683547
CAS Number: 55685-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR