3-(Benzyloxy)-6-bromo-2-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(Benzyloxy)-4-bromo-2-fluoro-3-methoxybenzene

36893-1G 36893-25G 36893-500MG 36893-5G
76888-702EA 236.59 USD
76888-702 76888-704 76888-706 76888-708
3-(Benzyloxy)-6-bromo-2-fluoroanisole ≥97%
3-(Benzyloxy)-6-bromo-2-fluoroanisole

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16618958
CAS Number: 1228957-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR