1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

36845-1G 36845-25G 36845-500MG 36845-5G
76888-690EA 379.81 USD
76888-690 76888-692 76888-694 76888-696
1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde ≥97%
1-(3-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carbaldehyde
Formula: C₁₁H₇F₃N₂O
MW: 240.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09951288
CAS Number: 75815-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR