1-(4-Bromophenyl)pyrrole ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(4-Bromophenyl)-1H-pyrrole
36834-25G
36834-5G
76888-678EA
310.85
USD
76888-678
76888-680
1-(4-Bromophenyl)pyrrole ≥97%
1-(4-Bromophenyl)pyrrole
Formula:
C₁₀H₈BrN MW: 222.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00128296 CAS Number: 5044-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...