(2-(Methoxymethyl)-4-(trifluoromethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

36813-1G 36813-25G 36813-500MG 36813-5G
76888-662EA 203.7 USD
76888-662 76888-664 76888-666 76888-668
(2-(Methoxymethyl)-4-(trifluoromethyl)phenyl)boronic acid ≥97% research grade
(2-(Methoxymethyl)-4-(trifluoromethyl)phenyl)boronic acid
Formula: C₉H₁₀BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632089
CAS Number: 2011780-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR