3-Nitrophenetole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Ethoxy-3-nitrobenzene

36801-25G 36801-5G
76888-650EA 254.62 USD
76888-650 76888-652
3-Nitrophenetole ≥95%
3-Nitrophenetole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024472
CAS Number: 621-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR