1-(4'-Methoxy-[1,1'-biphenyl]-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

36779-1G 36779-25G 36779-500MG 36779-5G
76888-620EA 195.21 USD
76888-620 76888-622 76888-624 76888-626
1-(4'-Methoxy-[1,1'-biphenyl]-4-yl)ethanone ≥95%
1-(4'-Methoxy-[1,1'-biphenyl]-4-yl)ethanone
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00438868
CAS Number: 13021-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR