1-Ethoxy-4-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36752-25G 36752-5G
76888-614EA 346.92 USD
76888-614 76888-616
1-Ethoxy-4-fluoro-2-methylbenzene ≥97%
1-Ethoxy-4-fluoro-2-methylbenzene
Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24237547
CAS Number: 1554472-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR