2-(Benzyloxy)-4-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36748-1G 36748-25G 36748-5G
76888-604EA 62.6 USD
76888-604 76888-606 76888-608
2-(Benzyloxy)-4-fluoro-1-methoxybenzene ≥97%
2-(Benzyloxy)-4-fluoro-1-methoxybenzene
Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18205778
CAS Number: 154051-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR