2,4-Dibromo-5-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

36736-1G 36736-25G 36736-500MG 36736-5G
76888-592EA 231.28 USD
76888-592 76888-594 76888-596 76888-598
2,4-Dibromo-5-methoxybenzaldehyde ≥95%
2,4-Dibromo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27935321
CAS Number: 99615-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR