2-(Benzo[d]thiazol-2-yl)-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

36638-1G 36638-25G 36638-5G
76888-568EA 62.6 USD
76888-568 76888-570 76888-572
2-(Benzo[d]thiazol-2-yl)-5-methylphenol ≥97%
2-(Benzo[d]thiazol-2-yl)-5-methylphenol
Formula: C₁₄H₁₁NOS
MW: 241.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043783
CAS Number: 56048-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR