1,4-Dibromo-2-chloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36567-1G 36567-25G 36567-5G
76888-518EA 109.28 USD
76888-518 76888-520 76888-522
1,4-Dibromo-2-chloro-3-methylbenzene ≥97%
1,4-Dibromo-2-chloro-3-methylbenzene
Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878178
CAS Number: 1000573-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR