(3-Bromo-2-Methylphenyl)(Methyl)Sulfane ≥97% research grade

Supplier: AOBChem USA

Warning

36548-1G 36548-25G 36548-500MG 36548-5G
76888-502EA 211.12 USD
76888-502 76888-504 76888-506 76888-508
(3-Bromo-2-Methylphenyl)(Methyl)Sulfane ≥97% research grade
(3-Bromo-2-Methylphenyl)(Methyl)Sulfane
Formula: C₈H₉BrS
Storage Temperature: Refrigerator
MDL Number: MFCD22488899
CAS Number: 871352-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR