1,3-Dibromo-5-iodobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
36526-25G
36526-5G
76888-490EA
204.76
USD
76888-490
76888-492
1,3-Dibromo-5-iodobenzene ≥97%
1,3-Dibromo-5-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃Br₂I MW: 361.80 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778996 CAS Number: 19752-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...