1-(4-Bromo-2,6-difluorophenyl)hexan-1-ol ≥95% research grade

Supplier: AOBChem USA

Warning

36509-25G 36509-5G
76888-478EA 524.09 USD
76888-478 76888-480
1-(4-Bromo-2,6-difluorophenyl)hexan-1-ol ≥95% research grade
1-(4-Bromo-2,6-difluorophenyl)hexan-1-ol
Formula: C₁₂H₁₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD32632021
CAS Number: 1598807-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR