5-Bromo-2,3,4-trifluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2,3,4-trifluorophenyl)methanol

36502-1G 36502-25G 36502-500MG 36502-5G
76888-466EA 173.99 USD
76888-466 76888-468 76888-470 76888-472
5-Bromo-2,3,4-trifluorobenzyl alcohol ≥97%
5-Bromo-2,3,4-trifluorobenzyl alcohol
Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756768
CAS Number: 511511-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR