2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(2-(Difluoromethoxy)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
36449-1G
36449-500MG
36449-5G
36449-25G
76888-420EA
498.63
USD
76888-420
76888-422
76888-424
77658-952
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester 95%
2-(Difluoromethoxy)-5-fluorobenzeneboronic acid, pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BF₃O₃ MW: 288.071 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30486443 CAS Number: 1417301-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...