2-(6-Chloropyridin-2-yl)acetic acid ≥97%

Supplier: AOBChem USA

Warning

36396-1G 36396-25G 36396-500MG 36396-5G
76888-408EA 290.69 USD
76888-408 76888-410 76888-412 76888-414
2-(6-Chloropyridin-2-yl)acetic acid ≥97%
2-(6-Chloropyridin-2-yl)acetic acid
Formula: C₇H₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08706288
CAS Number: 885267-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR