2,3,4-Trifluoroanisole ≥97%

Supplier: AOBChem USA

Warning

36372-25G 36372-5G
76888-380EA 286.45 USD
76888-380 76888-382
2,3,4-Trifluoroanisole ≥97%
2,3,4-Trifluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD00042396
CAS Number: 203245-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR