2-(Methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36326-1G 36326-25G 36326-5G
76888-344EA 70.02 USD
76888-344 76888-346 76888-348
2-(Methylthio)benzaldehyde ≥97%
2-(Methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈OS
MW: 152.2135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00196822
CAS Number: 7022-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR