(2-Bromo-6-fluoro-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

36321-1G 36321-25G 36321-500MG 36321-5G
76888-328EA 130.5 USD
76888-328 76888-330 76888-332 76888-334
(2-Bromo-6-fluoro-3-methoxyphenyl)methanol ≥97%
(2-Bromo-6-fluoro-3-methoxyphenyl)methanol
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26679560
CAS Number: 1500279-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR