3'-Bromo-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

36307-25G 36307-5G
76888-316EA 279.02 USD
76888-316 76888-318
3'-Bromo-2,2,2-trifluoroacetophenone ≥97%
3'-Bromo-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD00511232
CAS Number: 655-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR