2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethylphenylboronic acid pinacol ester

36265-25G 36265-5G
76888-286EA 183.54 USD
76888-286 76888-288
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₂
Storage Temperature: Ambient
MDL Number: MFCD18383605
CAS Number: 1232132-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR