1-(5-Bromopyridin-2-yl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

36195-1G 36195-25G 36195-500MG 36195-5G
76888-234EA 381.93 USD
76888-234 76888-236 76888-238 76888-240
1-(5-Bromopyridin-2-yl)-2,2,2-trifluoroethanol ≥97%
1-(5-Bromopyridin-2-yl)-2,2,2-trifluoroethanol
Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18909491
CAS Number: 917397-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR