1-Bromo-3,4-difluoro-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

36146-1G 36146-25G 36146-5G 36146-500MG
76888-220EA 99.73 USD
76888-220 76888-222 76888-224 77646-378
1-Bromo-3,4-difluoro-2-(methoxymethyl)benzene ≥97%
1-Bromo-3,4-difluoro-2-(methoxymethyl)benzene
Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707009
CAS Number: 2432848-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR