2,3-Difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

36034-25G 36034-5G
76888-178EA 215.37 USD
76888-178 76888-180
2,3-Difluorobenzamide ≥97%
2,3-Difluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061137
CAS Number: 18355-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR