1-Bromo-2-chloro-4-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

35919-1G 35919-25G 35919-5G
76888-166EA 65.78 USD
76888-166 76888-168 76888-170
1-Bromo-2-chloro-4-(difluoromethoxy)benzene ≥97%
1-Bromo-2-chloro-4-(difluoromethoxy)benzene
Formula: C₇H₄BrClF₂O
MW: 257.4599 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18390891
CAS Number: 345226-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR