1-(2-Bromo-4-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

35903-1G 35903-25G 35903-500MG 35903-5G
76888-154EA 376.62 USD
76888-154 76888-156 76888-158 76888-160
1-(2-Bromo-4-fluorophenyl)ethanol ≥97%
1-(2-Bromo-4-fluorophenyl)ethanol
Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
CAS Number: 261711-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR