1-Bromo-4-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,5-dimethyl-4-iodobenzene

35843-1G 35843-25G 35843-5G
76888-134EA 62.6 USD
76888-134 76888-136 76888-138
1-Bromo-4-iodo-2,5-dimethylbenzene ≥97%
1-Bromo-4-iodo-2,5-dimethylbenzene
Formula: C₈H₈BrI
MW: 310.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780644
CAS Number: 699119-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR