(2-(Methoxymethyl)phenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

35775-1G 35775-25G 35775-500MG 35775-5G
76888-108EA 379.81 USD
76888-108 76888-110 76888-112 76888-114
(2-(Methoxymethyl)phenyl)methanol ≥97% research grade
(2-(Methoxymethyl)phenyl)methanol
Formula: C₉H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18825431
CAS Number: 62172-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR