2-(Methoxymethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

35761-1G 35761-25G 35761-500MG 35761-5G
76888-092EA 124.13 USD
76888-092 76888-094 76888-096 76888-098
2-(Methoxymethyl)benzoic acid ≥97% research grade
2-(Methoxymethyl)benzoic acid
Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD09863839
CAS Number: 88550-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR